Learn More
CELLTREAT Scientific Products Inoculating Needle, Bulk Pack, Sterile
SDP
Supplier:  CELLTREAT Scientific Products 229615
Encompass_Preferred
CELLTREAT Scientific Products Inoculating Needle. Flexible for easy access to small volume tubes and dishes. Color coded yellow for easy identification. Gentle on delicate agar or other surfaces. 221mm Lenth. Made from polypropylene. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Bulk packed sterile, 20/pack, 2000/case.
Catalog No. 50-201-986