Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Inorganic Carbon Standard, 1000 ppm C, Ricca Chemical
SDP

Catalog No. 184516 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
500 mL
1 L
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
1845-16 500 mL Amber Glass Bottle
18454 120 mL Amber Glass Bottle
R18450001C 1 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 1845-16 Supplier Ricca Chemical Company Supplier No. 184516
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Chemical Identifiers

CAS 144-55-8 , 497-19-8 , 7732-18-5
Molecular Formula CHNaO3
Molecular Weight (g/mol) 84.01
MDL Number MFCD00003528
InChI Key UIIMBOGNXHQVGW-UHFFFAOYSA-M
IUPAC Name sodium hydrogen carbonate
SMILES [Na+].OC([O-])=O

Specifications

Packaging Amber Glass Bottle
Concentration or Composition Notes 1000ppm C
CAS Min % 97.0, 0.43, 0.34
CAS Max % 100.0, 0.45, 0.35
Equivalence 1 mL = 1 mg C
Physical Form Liquid
Color Colorless
Boiling Point 100°C
Quantity 500 mL
Chemical Name or Material Inorganic Carbon Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.