Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Walter Products™ Inorganic and Organic Chemistry Molecular Model Kit, 125 Pieces
SCI_ED

Catalog No. S139055 Shop All Walter Products Products
Change view
Click to view available options
Grade:
6+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S139055 6+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S139055 Supplier Walter Products™ Supplier No. MOLE1007
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Create colorful, visible structures to gain a better understanding of chemistry!

Build structural models of over 40 compounds!

  • Colored plastic spheres represent various atoms with holes at specific angles to accept plastic rods representing covalent and ionic bonds
  • Set includes 53 plastic spheres and 70 plastic connectors
  • Set includes a four compartment plastic storage box with a lid, disconnection tool and instruction manual

Specifications

For Use With (Application) Inorganic and Organic Chemistry
Grade 6+
Includes 125 Pieces
Material Plastic
Product Type Modeling Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.