Learn More
Invitrogen™ iNOS Monoclonal Antibody (CXNFT), Brilliant Violet™ 786, eBioscience™, Invitrogen™
Description
Specifications
Specifications
| Antigen | iNOS |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | CXNFT |
| Concentration | 0.2 mg/mL |
| Conjugate | Brilliant Violet 786 |
| Formulation | PBS with BSA and 0.09% sodium azide; pH 7.2 |
| Gene | Nos2 |
| Gene Accession No. | P29477 |
| Gene Alias | Hepatocyte NOS; hepatocytes; HEP-NOS; inducible nitric oxide synthase; inducible NO synthase; Inducible NOS; iNos; i-NOS; Inosl; MAC-NOS; macrophage NOS; nitric oxide synthase 2; nitric oxide synthase 2, inducible; nitric oxide synthase 2, inducible, macrophage; nitric oxide synthase 2A (inducible, hepatocytes); nitric oxide synthase, inducible; nitric oxide synthase, macrophage; nitric oxide synthase-inducible; NOS; NOS type II; NOS, type II; Nos2; Nos-2; Nos2a; NOS-II; OTTMUSP00000000202; peptidyl-cysteine S-nitrosylase NOS2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.