Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Inosine 5'-monophosphate, disodium salt hydrate, 97%

Catalog No. AC226260250
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC226260250 25 g Glass bottle
AC226260050 5 g Glass bottle
AC226261000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC226260250 Supplier Thermo Scientific Chemicals Supplier No. 226260250
Add to Cart

CAS: 352195-40-5 | C10H11N4Na2O8P | 392.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 352195-40-5
Molecular Formula C10H11N4Na2O8P
Molecular Weight (g/mol) 392.17
MDL Number MFCD00150372
InChI Key AANLCWYVVNBGEE-WCYUCLFNNA-L
Synonym inosine 5'-monophosphate disodium salt hydrate
PubChem CID 126963545
IUPAC Name disodium;[(2S,3S,4R,5R)-3,4-dihydroxy-5-(6-oxo-3H-purin-9-yl)oxolan-2-yl]methyl phosphate;hydrate
SMILES [Na+].[Na+].O[C@@H]1[C@@H](COP([O-])([O-])=O)O[C@H]([C@@H]1O)N1C=NC2=C1N=CNC2=O

Specifications

Melting Point 175.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 25 g
Beilstein 31, 26
Merck Index 14, 4977
Solubility Information Solubility in water: soluble
Spectrophotometry 96% min. (lambda max. 249nm,pH 7) molar absorptivity 12300+/-4%,on dry basis
Formula Weight 392.16
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Inosine 5'-monophosphate, disodium salt hydrate
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement:
May cause damage to organs through prolonged or repeated exposure.
Suspected of damaging fertility.
Precautionary Statement GHS P Statement:
Use personal protective equipment as required.
Do not breathe dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.