Learn More
U.S. Pharmacopeia Inositol, 87-89-8, MFCD00077932, 500mg
Supplier:  U.S. Pharmacopeia 1340960
Encompass
Molecular formula C6H12O6, Molecular weight 180.16, Melting Point 435.2 - 440.6 °F (224 - 227 °C), Synonyms: Myo-inositol
Catalog No. NC0189755
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.