Learn More
BioMed Diagnostics A self-contained system for the detection of Trichomonas gallinae/Tetratrichomonas gallinarum from avian specimens. | Available in 100 pack
Supplier:  BioMed Diagnostics 12601002
Encompass_Preferred
A self-contained system for the detection of Trichomonas gallinae/Tetratrichomonas gallinarum from avian specimens. The proprietary medium is highly selective for T.gallinae/T.gallinarum and inhibits the growth of other organisms that can interfere with a reliable diagnosis. | Detects: Trichomonas gallinae/Tetratrichomonas gallinarum | Available in: 100 pack
Catalog No. NC2582893
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.