Learn More
Echelon Biosciences Research Labs Ins(1,3,4,5)P4 100 ug
SDP
Supplier:  Echelon Biosciences Research Labs Q1345100UG
Encompass
Inositol phosphates are important signaling molecules involved in numerous cellular functions. Inositol 1,3,4,5-tetrakisphosphate (Ins(1,3,4,5)P4), aka IP4, is produced from Ins(1,4,5)P3 in stimulated cells by IP3 3-kinase (Itpkb). IP4 acts in conjunction with IP3 to mobilize intracellular calcium. In addition, IP4 has roles in T cell precursor development and responsiveness and thymocyte reactivity to antigens. This item is non returnable
Catalog No. 50-720-734
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.