Learn More
United Scientific™ Insect Pins with Yellow Epoxy Head
SDPSCI_ED
Catalog No. S04536A Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Size:
0
00
000
1
2
3
4
5
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
S04536A 0
S04536 00
S04536C 2
S04536B 1
S04536D 3
S04536E 4
S04536F 5
S15915 000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. S04536A Supplier United Scientific™ Supplier No. IPINS0PK/100
Add to Cart
Add to Cart
Insect Pins with Yellow Epoxy Head are coated with a black corrosion-resistant finish.
  • Steel pins
  • Yellow epoxy heads
  • Available in sizes 000 through 5
Specifications
Product Type Insect Pins
Autoclavable Not Autoclavable
Coating Black Corrosion Resistant
Material Steel
For Use With (Application) Dissection
Includes Yellow epoxy head
Size 0