Learn More
United Scientific™ Insect Pins with Yellow Epoxy Head
SDPSCI_ED
Insect Pins with Yellow Epoxy Head are coated with a black corrosion-resistant finish.
Supplier:  United Scientific™ IPIN04PK/100
 View more versions of this product
Catalog No. S04536E
Only null left
Add to Cart
Description
Description
  • Steel pins
  • Yellow epoxy heads
  • Available in sizes 000 through 5
Specifications
Specifications
Insect Pins
Black Corrosion Resistant
Dissection
4
Not Autoclavable
Steel
Yellow epoxy head
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.