Learn More
Perkin Elmer US LLC Sealing Insert for Standard Aluminum Pans
Supplier:  Perkin Elmer US LLC B0508921
Encompass_Preferred
The Universal Crimper Press requires Insert (B0144637) for Volatile Sample Pans (02190062, 02190080, and N5190788), Insert (B0505340) for Stainless Steel Pans (03190218 and 03190029), Insert (B0508921) for Standard Aluminum Pans (02190041), and Insert (B0139033) for hermetically-sealed 10, 30, and 50 µL sample pans.
Catalog No. 50-904-2512