Learn More
Mopec Inside Corner Filler, Standing Height, Adjustable
SDP
Catalog No. 2123671 Shop All Mopec Products
Change view
Click to view available options
Product Type:
Corner Filler
Catalog No. Product Type
21-236-71 Corner Filler
1 options
Catalog No. 21-236-71 Supplier Mopec Supplier No. LCFCHADJS
Add to Cart
Add to Cart
Mopec’s Casework Line offers durable, modular stainless-steel cabinetry for surgical pathology, autopsy, and lab settings. Built for longevity, easy cleaning, and seamless integration, it supports efficient, organized, and ergonomic workflows in clinical environments.
Construction
  • (1) Standing height inside corner filler, adjustable
  • Fillers help create a seamless and polished look, ensuring that the cabinetry appears cohesive and well-integrated into the design of the room.
Materials
  • 304 stainless steel construction with a polished No. 4 finish
Dimensions
  • LCFCH-ADLC: 18 in. L x 18 in. W x 34.6 in. H
Specifications
Product Type Corner Filler