Learn More
Thermo Scientific™ InSight Monitoring Kit (T/RH+)
Catalog No. 11676384 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Incubators
Catalog No. For Use With (Equipment)
11-676-384 Incubators
1 options
Catalog No. 11-676-384 Supplier Thermo Scientific™ Supplier No. INSKTINCU104
This product is not available for sale online. Please call Customer Service at 1-800-766-7000 or send an email to cs.webemail@thermofisher.com for assistance.
InSight Asset management solution for Laboratory Equipment enables knowledge based decisisons by monitoring critical equipment parameters, providing data insights and various asset manegement tools.
Thermo Scientific™ InSight is a wireless monitoring and Asset management solution that measures critical realiability parameters and uses advanced analytics to provide pedictive indicators of possible system failures. InSight consists of wireless modules (motes), sensors and software tools that allow you to monitor and manage your laboratory equipment. Alarms are issued automatically to users’ email and mobile devices when setpoint breaches are detected in the system.
InSight Analytics:
InSight Analytics, powered by KLATU™, is a system designed to collect, analyze and make efficiency and operating cost recommendations from multiple asset inputs and events such as:
  • Equipment critical parameters
  • Electrical use and energy consumption
  • Asset utilization
  • Environmental conditions
InSight Analytics monitors your laboratory equipment 24 hours a day, seven days a week and alerts you when the thresholds are exceeded. The data from InSight Analytics is run through a number of algorithms utilizing the InSight software. With InSight Analytics, you can determine and compare the assets' performance against industry performance standards.
How InSight Analytics works:
  • Data is sent wirelessly from the InSight mote via your Wi-Fi network to the InSight web platform fpr storage.
  • Once in the Cloud, the data is analyzed by the system, providing real-time comparison of the laboratory equipment's health to industry averages.
  • This data analysis enables you to make knowledge-based asset decisions based on key performance indicators such as product service and support, service vs. buy decisions and remote system health monitoring and trouble-shooting for service providers.
TRUSTED_SUSTAINABILITY
Specifications
Type Accessory, Monitoring Kit
Description InSight Kit for Incubator (Temp/RH Condensable)
For Use With (Equipment) Incubators