Learn More
Thermo Scientific™ InSight Kits
Catalog No. 11676377
Change view
Click to view available options
For Use With (Equipment):
LN2 Cryo Storage
ULT Freezers, Freezers, Refrigerators
Walk-in w/ Dual Refrigeration System
Walk-in w/ Single Refrigeration System
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
11-676-377 ULT Freezers, Freezers, Refrigerators
11-676-380 LN2 Cryo Storage
11-676-378 Walk-in w/ Single Refrigeration System
11-676-379 Walk-in w/ Dual Refrigeration System
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 11-676-377 Supplier Thermo Scientific™ Supplier No. INSKTCOLD101
This product is not available for sales online. Please call Customer Service at 1-800-766-7000 or send an email to cs.webemail@thermofisher.com for assistance.
Proactively monitor your cold storage equipment’s temperature, energy consumption, and other parameters using Wi-Fi or proprietary wireless protocols.
Thermo Scientific™ InSight is a wireless monitoring solution that has been designed to proactively monitor your cold storage equipment. InSight consists of wireless modules (motes), sensors and software tools that allow you to monitor your cold storage equipment’s temperature, energy consumption, door access, reliability, predictive failure analytics, as well as other parameters using Wi-Fi or proprietary wireless protocols. Alarms are issued automatically to users’ email and mobile devices when setpoint breaches are detected in the system.
InSight Analytics:
InSight Analytics, powered by KLATU™, is a system designed to collect, analyze and make efficiency and operating cost recommendations from multiple asset inputs and events such as:
  • Equipment critical parameters
  • Electrical use and energy consumption
  • Asset utilization
  • Environmental conditions
InSight Analytics monitors your laboratory equipment 24 hours a day, seven days a week and alerts you when the thresholds are exceeded. The data from InSight Analytics is run through a number of algorithms utilizing the InSight software. With InSight Analytics, you can determine and compare the assets' performance against industry performance standards.
How InSight Analytics works:
  • Data is sent wirelessly from the InSight mote via your Wi-Fi network to the InSight web platform fpr storage.
  • Once in the Cloud, the data is analyzed by the system, providing real-time comparison of the laboratory equipment's health to industry averages.
  • This data analysis enables you to make knowledge-based asset decisions based on key performance indicators such as product service and support, service vs. buy decisions and remote system health monitoring and trouble-shooting for service providers.
TRUSTED_SUSTAINABILITY
Specifications
Type Insight Monitoring Kit
Display Type No Display
Calibration Not Calibrated
Alarm Types Mobile Device
Software Type Insightlink Gateway
Description InSight Kit for Ultra-Low Temperature and Freezers
For Use With (Equipment) ULT Freezers, Freezers, Refrigerators
Not available outside North America