Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Akro-Mils™ InSight™ Storage System: 10 lb., 53/8 x 4/18 x 31/4 in.

Catalog No. 08757674
Encompass
Encompass_Preferred
Change view
Click to view available options
Type:
Bin Lid
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
08-757-674 Bin Lid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 08-757-674 Supplier Akro-Mils™ Supplier No. 305A2
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Visible organization solutions for healthcare and industrial workspaces

InSight bins offer transparent storage with a sleek, modern design. Can be stacked, shelved or hung on Akro-Mils rails and louvered panels or ReadySpace™ Storage System.

  • Constructed of durable polycarbonate
  • Autoclavable
  • No tip feature for secure stacking
  • Rear lip supports bin on hanging systems
  • Large label area for easy identification and bar code scanning
  • Optional lids and length/width dividers available to create dust-free, custom-size compartments

Specifications

Type Bin Lid
Material Polycarbonate
Color Clear
Autoclavable Autoclavable
For Use With (Equipment) 305A1 Bin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.