Learn More
Description
- No reagents or field kits required for most strips
- Chemically treated pads on plastic strips produce color change
- Pop-top vials have molded desiccant liner and hinged snap cap to protect from moisture
- As little as 10-second development time
- Test strips for testing water: wastewater, industrial water boiling and cooling, drinking water, laundry and sanitation †
Specifications
Specifications
| For Use With (Application) | Water quality testing |
| Parameter Specifications | Copper |
| Quantity | 25 Strips/Vial |
| Range | 0-0.3-0.6-1-3 ppm |
| Storage Requirements | Store tightly sealed vial in a cool, dry storage area away from moisture; Keep out of direct sunlight |
| Type | Single Factor Test Strip |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

