Description
The InstaFlux™ Barcode Reader is an accessory designed to simplify sample number entry and optimize data management in the lab. It enables fast and error-free input, reducing the risk of inconsistencies. Compatible with LIMS systems, it ensures smooth and automated traceability of analyses.
Features and Key Benefits
- Smooth integration
- Enhanced traceability
- Reliable sample identification
- Ease of use
- 5-year shelf life
Specifications
Specifications
| Supported Barcode Types | 1D linear barcodes and 2D Data Matrix / QR codes |
| For Use With (Equipment) | InstaDose™ Gravimetric Diluter, InstaFill&trade Peristaltic Pump |
| Type | Barcode Reader |
| Power Supply | USB |
| Plug Type | USB |