Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Enhancer Solution

Catalog No. 50246702
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-246-702 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-246-702 Supplier Invitrogen™ Supplier No. IOES1
  • $157.00
    $152.65 / Each of 1

    Save $4.35

    3% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Added to the 5x Cell Lysis Buffer (Cat. No. IOCLB1) to yield the Cell Lysis Mix solution that is applied to cultured cells or tissues prior to running the assays.

  • The 5x Cell Lysis Mix is used to lyse cells in the presence of culture medium and is typically used to lyse non-adherent cells
  • Dilute 5x Cell Lysis Mix 5-fold with water to make a 1x Cell Lysis Mix
  • 1x Mix is used to lyse cells after the removal of culture medium, and is typically used to lyse adherent cells or non-adherent cells that have been harvested by centrifugation
  • Lysis Mix should be used as the diluent for any dilution of cellular lysates that are required
  • Supplementing Cell Lysis Mix with extra components (e.g. protease inhibitors, chelating agents, detergents) should be tested on a case-by-case basis for compatibility with InstantOne ELISA assays
TRUSTED_SUSTAINABILITY

Specifications

Form Liquid
For Use With (Application) ELISA
Quantity 100 mL
Reagent Type Signal Enchancers
Storage Requirements 2°C to 8°C
Validated Application ELISA
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.