Learn More
Sartorius INSTRUMENT OCTET-RH16 30-5101
Catalog No. 502891661 Shop All Sartorius Products
Encompass
Octet RH16 is a label-free biomolecular interaction analysis platform that facilitates rapid characterization and development of biologics drug molecules. It provides high detection sensitivity and is compatible with 384-microwell plates making it ideal for small molecule library screening and large cross-competition binding analysis. The platform is highly automated reducing operator input and extending walk-away times. High Detection Sensitivity and Throughput: The Octet RH16 system is ideal for small molecule library screening applications requiring high sensitivity throughput and sample capacity. Advanced Automation and Efficiency: The system is compatible with external automation to reduce operator input and increase efficiency with automation-ready and robot-compatible features for microplate and biosensor tray loading. Ligands can be immobilized in batches onto biosensors for future experiments maximizing flexibility and cost-effectiveness.
Catalog No. 50-289-1661 Supplier Sartorius Supplier No. OCTETRH16
To request a quote for this item, please select Request a Quote to reach our dedicated support team.
Octet RH16 is a label-free biomolecular interaction analysis platform that facilitates rapid characterization and development of biologics drug molecules. It provides high detection sensitivity and is compatible with 384-microwell plates making it ideal for small molecule library screening and large cross-competition binding analysis. The platform is highly automated reducing operator input and extending walk-away times. High Detection Sensitivity and Throughput: The Octet RH16 system is ideal for small molecule library screening applications requiring high sensitivity throughput and sample capacity. Advanced Automation and Efficiency: The system is compatible with external automation to reduce operator input and increase efficiency with automation-ready and robot-compatible features for microplate and biosensor tray loading. Ligands can be immobilized in batches onto biosensors for future experiments maximizing flexibility and cost-effectiveness.