Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SHOWA™ PVC-Coated Triple Insulated Gloves - Reinforced Gauntlet Cuff

Catalog No. 19014978 Shop All Showa Best Glove Products
Change view
Click to view available options
Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-014978 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-014978 Supplier SHOWA™ Supplier No. 7510
This item is not returnable. View return policy

Three layers of foam insulation provide protection against chemicals, and cold weather. PVC-coating has a rough finish for enhanced grip capabilities in wet or dry conditions.

  • Three layers of foam insulation between a layer of cotton-jersey and cotton interlock for increased cold-weather protection
  • Low-temperature compound remains flexible, even in sub-zero temperatures
  • Reinforced gauntlet extends coverage to wrist and forearm
  • PVC-coating provides abrasion and chemical resistance
  • Rough finish enhances grip capabilities in wet and dry conditions

Specifications

Cuff Material Reinforced Cotton
Coating Material PVC
Color Orange
Exterior Finish Fully Textured
Hand Specific
Material Cotton
Size Large
For Use With (Application) Signalers, Digging, Utility Work, and Heavy Equipment Handling
Glove Cuff Style Gauntlet
Liner Material Cotton Jersey
Series 75
Length (English) 12 in.
Length (Metric) 30.5 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.