Learn More
Creative Biomart Insulin Pork(Native purified)
Supplier:  Creative Biomart THP0131
Encompass_Preferred
Insulin isolated from pig pancreas. Composed of alpha and beta chains, processed from pro-insulin. Forms a hexameric structure.
Catalog No. 50-208-4182
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.