Learn More
Arbor Assays Insulin ELISA Kit (1 Plate)
SDP
Supplier:  Arbor Assays K046H1
Encompass_Preferred
The DetectX Insulin ELISA Kit is designed to quantitatively measure insulin present in a variety of samples and tissue culture media. 
Catalog No. 50-227-8456
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.