Learn More
Crystal Chem Inc Insulin ELISA Kit (10 Kit Pack), 96-wells per kit, Individually packaged
SDP
Supplier:  Crystal Chem Inc 90096
Encompass
The Insulin ELISA 10-kit pack is an economical option for high-volume users of our Insulin ELISA. Each kit is idividually packaged for ease of use. Highlights: ~100% Cross-reactivity to insulin and most analogs. Very low (1.2%) reactivity to proinsulin. Very low (< 5%) reactivity to mouse/rat insulin. Fast assay time of 2 hrs 15 mins. Wide dyanamic range of 0.9 - 220 mU/L.Crystal Chem is a worldwide leading supplier of ELISA kits for human markers. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7923
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.