Learn More
Tocris Bioscience™ Insulin (human) recombinant, expressed in yeast
GREENER_CHOICE
Catalog No. 343510
Change view
Click to view available options
Quantity:
10 mg
Catalog No. Quantity
34-351-0 10 mg
1 options
This item is not returnable. View return policy
Catalog No. 34-351-0 Supplier Tocris Bioscience™ Supplier No. 3435/10
Add to Cart
Add to Cart
This item is not returnable. View return policy
Endogenous peptide agonist
Insulin (human, recombinant, expressed in yeast) is an endogenous insulin receptor agonist (Ki = 4.85 nM). Decreases plasma glucose levels, proteolysis, lipolysis and gluconeogenesis and increases glycogen and fatty acid synthesis in vivo.
Specifications
Quantity 10 mg