Learn More
BioVendor, LLC Insulin Multispecies (human, rabbit, dog, monkey) ELISA (96 wells)
SDP
Supplier:  BioVendor, LLC YK060
Encompass
Insulin Multispecies (human, rabbit, dog, monkey) ELISA (96 wells). Enzyme immunoassay kit for quantitative determination of insulin in serum sample of human, rabbit, dog/canine, and monkey. Sensitive quantification, high specificity, and no influences with other components in samples. No sample pretreatment needed. Recombinant human insulin standard. SAMPLE REQ.: 25 l/well. ASSAY RANGE: 0.137-100 ng/mL. SUMMARY: This kit is a stable and convenient assay system for measurement of insulin (human, rabbit, monkey, and dog) in serum samples. The processing of proinsulin, which occurs within the B cell, yields insulin and C-peptide. Insulin and C-peptide are secreted in equimolar quantities into blood circulation. Therefore, the measurement of insulin in blood is very important, and also provides valuable information to evaluate the pancreatic B cell function. Manufactured by Yanaihara Institute Inc., Japan.
Catalog No. 50-421-6
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.