Learn More
Alpco Ltd Rat Ultrasensitive Insulin ELISA
SDP
Supplier:  Alpco Ltd 80INSRTUE01
Encompass_Preferred
The Rat Ultrasensitive Insulin ELISA quantifies the concentration of insulin protein products in rat I and rat II proinsulin genes from as little as 5 µL of serum or plasma sample. For Research Use Only. Not for Use in Diagnostic Procedures.
  • Only 5 µL of sample required for tail snips
  • Dynamic range of 0.15-5.5 ng/mL
  • Superior sensitivity of 0.107 ng/mL
  • Only 25 µL of sample required for hypoinsulinemic samples
  • Dynamic range of 0.02-1.0 ng/mL
  • Superior sensitivity of 0.002 ng/mL
Catalog No. 50-922-596
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.