Learn More
Sino Biological Human Insulin Receptor / INSR / CD220 Protein (long isoform, His Tag), Biotinylated 100ug
Supplier:  Sino Biological 11081H08HB100
Encompass_Preferred
A DNA sequence encoding the human INSR isoform long (NP_000199.2) extracellular domain (Met1-Lys956) was expressed, fused with a polyhistidine tag at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-211-6115
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.