Learn More
Supply Solutions Insulin solution from bovine pancreas, MFCD00131380, 5mL
Supplier:  Supply Solutions SIGMA I05165ML
Encompass_Preferred
10 mg/mL insulin in 25 mM HEPES, pH 8.2, BioReagent, sterile-filtered, suitable for cell culture
Catalog No. NC0829811
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.