Learn More
TARGETMOL CHEMICALS INC INSULINCATTLE 50MG
SDP
Supplier:  TARGETMOL CHEMICALS INC TP1125
Encompass_Preferred
Also available in 10 mg 25 mg 100 mg 200 mg and bulk. Please contact Fisher for quotes. Insulin cattle (Insulin from bovine pancreas) is a two-chain polypeptide hormone produced in vivo in the pancreatic (beta) cells。Ithas often been used as growth supplement in culturing cells. purity: 98%
Catalog No. 50-257-6165
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.