Learn More
ACROBiosystems ACROBiosystems Human Insulin R / CD220 (28-944) Protein, His Tag (SPR verified)
Supplier:  ACROBiosystems INRH52HA100UG
Encompass_Preferred
Recombinant Protein;100UG;Human Insulin R (28-944), His Tag (SPR verified) (INR-H52Ha) is expressed from human 293 cells (HEK293). It contains AA His 28 - Lys 944 (Accession # P06213-2).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 106.5 kDa. The protein migrates as 115-125 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human Insulin R (28-944), His Tag (Cat. No. INR-H52Ha) at 5 μg/mL (100 μL/well)can bind Human IGF-II, Fc Tag (Cat. No. IG2-H4260) with a linear range of 0.078-1.25 μg/mL (QC tested).;Immobilized Human Insulin R (28-944), His Tag (Cat. No. INR-H52Ha) at 5 μg/mL (100 μL/well)can bind Insulin Biotin Conjugate with a linear range of 0.02-0.313 μg/mL (Routinely tested).;Human Insulin R (28-944), His Tag (Cat. No. INR-H52Ha) captured on CM5 chip via anti-His antibody, can bind Human Insulin with an affinity constant of 249 nM as determined in a SPR assay (Biacore T200) (QC tested).
Catalog No. 50-195-7001
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.