Learn More
Crystal Chem Inc Intact Proinsulin Chemiluminescence Kit, 5 Kit Pack, 96-wells per kit
SDP
Supplier:  Crystal Chem Inc 90107
Encompass_Preferred
The Intact Proinsulin Chemiluminescence is a highly sensitive assay that is used to quantify levels of human intact proinsulin in serum and plasma. Highlights: Kits use a sample volume of only 50 µL. Highly specific for intact proinsulin. Dynamic range of 0.02 - 100 pmol/mL. Highly precise with low CV's. Crystal Chem is a worldwide leading supplier of ELISA kits for human markers. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7925
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.