Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integra™ Miltex™ Mayo Scissors

Catalog No. 12460466
Change view
Click to view available options
Length (English):
5.5 in.
6.75 in.
9 in.
Blade Style:
Curved, Beveled
Straight, Beveled
Straight, Rounded Tips
Product Type:
Carb-N-Sert Mayo Scissors
Mayo Scissors
SuperCut Mayo Scissors
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English) Blade Style Product Type
12-460-466 5.5 in. Straight, Beveled Mayo Scissors
12-460-467 5.5 in. Curved, Beveled Mayo Scissors
12-460-468 5.5 in. Curved, Beveled Carb-N-Sert Mayo Scissors
12-460-471 6.75 in. Curved, Beveled Mayo Scissors
12-460-469 6.75 in. Straight, Beveled Mayo Scissors
12-460-470 6.75 in. Straight, Beveled Carb-N-Sert Mayo Scissors
12-460-472 9 in. Straight, Beveled Mayo Scissors
12-460-473 5.5 in. Straight, Rounded Tips Mayo Scissors
12-460-474 5.5 in. Straight, Rounded Tips Carb-N-Sert Mayo Scissors
12-460-475 6.75 in. Straight, Rounded Tips Carb-N-Sert Mayo Scissors
12-460-506 5.5 in. Straight, Beveled SuperCut Mayo Scissors
12-460-508 6.75 in. Straight, Beveled SuperCut Mayo Scissors
12-460-507 5.5 in. Curved, Beveled SuperCut Mayo Scissors
12-460-509 9 in. Straight, Beveled SuperCut Mayo Scissors
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Catalog No. 12-460-466 Supplier Integra™ Miltex™ Supplier No. 5120
Add to Cart
Add to Cart

Offer outstanding distal cutting capability

  • High-quality surgical-grade stainless steel construction provides optimal performance
  • Fully tempered precision screw locks maintain perfect alignment
  • Ergonomic design combines weight, balance and feel for optimal comfort and performance
  • Well rounded and polished ring handles provide exceptional comfort and reduced hand fatigue
  • Sharp/sharp scissors available in straight and curved patterns
  • Carb-N-Sert™ scissors have tungsten carbide inserts seamlessly bonded to upper and lower blades and feature gold ring handles
  • SuperCut scissors have a specially designed razor sharp edge that cuts effortlessly through tissue; lower blade has micro fine serrations to prevent tissue slippage

Order Info

Cold or chemical sterilization not recommended for Carb-N-Sert scissors. SuperCut scissiors designed to cut tissue only; not recommended for use on suture material. Dropping or mishandling will cause the razor-shapr edge to nick or become irregular.

Specifications

Product Type Mayo Scissors
Blade Style Straight, Beveled
Grade Premium
Length (English) 5.5 in.
Length (Metric) 13.97 cm
Material Stainless Steel
For Use With (Application) Dissecting
Tip Style Blunt/Blunt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.