Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integra™ Miltex™ Metzenbaum Scissors

Catalog No. 12460480
Change view
Click to view available options
Length (Metric):
11.43 cm
12.7 cm
13.97 cm
17.78 cm
22.86 cm
Blade Style:
Curved
Straight
Product Type:
Carb-N-Sert Metzenbaum Scissors
Metzenbaum Scissors
SuperCut Metzenbaum Scissors
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Blade Style Product Type
12-460-480 13.97 cm Curved Metzenbaum Scissors
12-460-482 17.78 cm Curved Metzenbaum Scissors
12-460-479 13.97 cm Straight Carb-N-Sert Metzenbaum Scissors
12-460-481 17.78 cm Straight Metzenbaum Scissors
12-460-483 22.86 cm Straight Metzenbaum Scissors
12-460-488 12.7 cm Curved Metzenbaum Scissors
12-460-487 12.7 cm Straight Metzenbaum Scissors
12-460-511 13.97 cm Curved SuperCut Metzenbaum Scissors
12-460-513 11.43 cm Curved SuperCut Metzenbaum Scissors
12-460-512 11.43 cm Straight SuperCut Metzenbaum Scissors
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 12-460-480 Supplier Integra™ Miltex™ Supplier No. 5180
Add to Cart
Add to Cart

Precisely cuts tough or delicate tissue

  • High-quality surgical-grade stainless steel construction provides optimal performance
  • Fully tempered precision screw locks maintain perfect alignment
  • Ergonomic design combines weight, balance and feel for optimal comfort and performance
  • Well rounded and polished gold ring handles provide exceptional comfort and reduced hand fatigue
  • Straight or curved blades
  • Blunt points
  • Tungsten carbide inserts seamlessly bonded to upper and lower blades
  • SuperCut scissors have a specially designed razor sharp edge that cuts effortlessly through tissue; lower blade has micro fine serrations to prevent tissue slippage

Order Info

Cold or chemical sterilization not recommended for tungsten carbide scissors. SuperCut scissiors designed to cut tissue only; not recommended for use on suture material. Dropping or mishandling will cause the razor-shapr edge to nick or become irregular.

Specifications

Product Type Metzenbaum Scissors
Blade Style Curved
Grade Premium
Length (English) 5.5 in.
Length (Metric) 13.97 cm
Material Stainless Steel
For Use With (Application) Surgical, metzenbaum
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.