Learn More
Integra™ Miltex™ Micro Alligator Ear Forceps
Catalog No. 12460336
Change view
Click to view available options
Width (Metric):
0.5 mm
0.6 mm
0.8 mm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Width (Metric)
12-460-336 0.8 mm
12-460-335 0.5 mm
12-460-334 0.6 mm
12-460-337 0.8 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 12-460-336 Supplier Integra™ Miltex™ Supplier No. 192100
Add to Cart
Add to Cart
Delicate design for foreign body removal
  • Precision grasping and extraction of cerumen from the ear canal
  • Satin-finish stainless steel
  • Autoclavable
Specifications
Product Type Forceps, Micro, Alligator Ear
Finish Satin
For Use With (Application) Foreign body removal
Quantity Each
Style Micro Alligator Ear
Tip Style Serrated
Width (Metric) 0.8 mm