Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integra™ Miltex™ Operating Room Scissors

Catalog No. 12460502
Change view
Click to view available options
Length (English):
4.5 in.
5 in.
5.5 in.
6.5 in.
7.5 in.
Blade Style:
Curved
Straight
Straight, Supercut
Material:
Stainless Steel
Tungsten Carbide
Tip Style:
Blunt/Blunt
Sharp/Blunt
Sharp/Sharp
17 product options available for selection
Product selection table with 17 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English) Blade Style Material Tip Style
12-460-502 5.5 in. Straight Tungsten Carbide Sharp/Sharp
12-460-103 5.5 in. Straight Stainless Steel Sharp/Blunt
12-460-107 5.5 in. Straight Stainless Steel Sharp/Sharp
12-460-113 4.5 in. Curved Stainless Steel Sharp/Sharp
12-460-114 5.5 in. Curved Stainless Steel Sharp/Sharp
12-460-130 5.5 in. Curved Stainless Steel Blunt/Blunt
12-460-148 5 in. Straight Stainless Steel Sharp/Sharp
12-460-147 4.5 in. Straight Stainless Steel Sharp/Sharp
12-460-150 4.5 in. Straight Stainless Steel Sharp/Blunt
12-460-152 7.5 in. Straight Stainless Steel Sharp/Blunt
12-460-149 6.5 in. Straight Stainless Steel Sharp/Sharp
12-460-153 4.5 in. Straight Stainless Steel Blunt/Blunt
12-460-151 5 in. Straight Stainless Steel Sharp/Blunt
12-460-154 5.5 in. Straight Stainless Steel Blunt/Blunt
12-460-155 6.5 in. Straight Stainless Steel Blunt/Blunt
12-460-478 5.5 in. Straight Tungsten Carbide Sharp/Blunt
12-460-510 5.5 in. Straight, Supercut Stainless Steel Sharp/Blunt
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 17 options available.
17 options
Catalog No. 12-460-502 Supplier Integra™ Miltex™ Supplier No. 56TC
Add to Cart
Add to Cart

For general utility applications

  • High-quality surgical-grade stainless steel construction provides optimal performance
  • Fully tempered precision screw locks maintain perfect alignment
  • Ergonomic design combines weight, balance and feel for optimal comfort and performance
  • Well rounded and polished ring handles provide exceptional comfort and reduced hand fatigue
  • Standard pattern
  • Blunt-blunt and sharp-blunt tips are ideal for cutting sutures, gauze, tubing and other materials
  • Sharp-sharp tips are preferred for cutting tendons and cartilage
  • SuperCut scissors have a specially designed razor sharp edge that cuts effortlessly through tissue; lower blade has micro fine serrations to prevent tissue slippage

Order Info

SuperCut scissiors designed to cut tissue only; not recommended for use on suture, gauze or tubing. Dropping or mishandling will cause the razor-shapr edge to nick or become irregular.

Specifications

Product Type Surgical Scissors
Blade Style Straight
Grade Premium
Length (English) 5.5 in.
Length (Metric) 13.97 cm
Material Tungsten Carbide
For Use With (Application) Surgical, Precision, Carb-N-Sert
Tip Style Sharp/Sharp
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.