Learn More
Integra™ Miltex™ Sterile Surgical Blades
Catalog No. 12460443
Change view
Click to view available options
Size:
# 10
# 11
# 12
# 15
# 21
# 22
# 25
25.5 cm
Blade Material:
Carbon Steel
Stainless Steel
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Blade Material
12-460-443 # 15 Carbon Steel
12-460-108 25.5 cm Stainless Steel
12-460-441 # 11 Carbon Steel
12-460-440 # 10 Carbon Steel
12-460-442 # 12 Carbon Steel
12-460-444 # 21 Carbon Steel
12-460-445 # 22 Carbon Steel
12-460-449 # 22 Stainless Steel
12-460-447 # 11 Stainless Steel
12-460-448 # 15 Stainless Steel
12-460-446 # 25 Carbon Steel
12-460-450 # 25 Stainless Steel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. 12-460-443 Supplier Integra™ Miltex™ Supplier No. 4115C
Add to Cart
Add to Cart
Single-use blades for convenience
  • Sterile
  • Razor-sharp cutting edge
  • Sizes clearly marked on each blade
  • Individually packaged
Specifications
Blade Material Carbon Steel
Product Type Surgical Blade
For Use With (Application) For Superior Cutting and Clean Incisions
Size # 15
For Use With (Equipment) For # 3 or # 4 Scalpel Handles
Disposable Single-use
Sterility Sterile