Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poltex™ Integra Pipette Rack 6-position Mount
SDP

Catalog No. 22000804
Change view
Click to view available options
Mounting:
VHB Tape
Wall Mount
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mounting
22-000-804 VHB Tape
22-000-816 Wall Mount
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-000-804 Supplier Poltex™ Supplier No. INTEG6T
Add to Cart
Add to Cart

Mount for Pipetter or Multichannel Hanger for Integra™ or similarly styled pipetters.

  • PETG construction
  • Fits up to 6 pipetters
  • Choose from wall mount or VHB tape

Specifications

Autoclavable Not Autoclavable
Color Clear
Holds Pipets
Material PETG
Positions 6
Shape Rectangular
Stackable No
Sterility Non-sterile
For Use With (Equipment) Integra Pipetter
Mounting VHB Tape
Size Small
Length (Metric) 127 mm
Product Type Pipet Rack
Shelves None
Height (Metric) 106 mm
Array 1 x 6
Width (Metric) 330 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.