Description
- Available in a wide variety of frame colors and lens tints
- Select frames available with fog free coating
- Lenses are coated for superior scratch resistance
- Lenses provide 99% protection against harmful UV rays
Specifications
Specifications
| Lens Coating | Scratch-resistant |
| Lens Tint | Clear |
| Product Type | Safety Glasses |
| For Use With (Application) | Eye protection |
| Frame Color | Red |
| Temple Style | Adjustable |
| Lens Material | Polycarbonate |
| Frame Type | Classic |