Learn More
Promega Integrated Reagent Caps for ReliaPrep™ Large Volume HT gDNA Isolation System
Catalog No. PRA2701 Shop All Promega Corporation Products
Encompass
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
4/Pk.
Catalog No. Quantity
PRA2701 4/Pk.
1 missing translation for 'options'
Catalog No. PRA2701 missing translation for 'mfr' Promega missing translation for 'supplierNo' A2701
Add to Cart
Add to Cart
Accessory for use with the ReliaPrep Large Volume HT gDNA System used in automated purification of DNA from 1-10 mL blood on liquid-handling systems.
  • Accessory for use with the ReliaPrep™ Large Volume HT gDNA Isolation System
  • Reagent caps used with reagents for automated purification of DNA from 1-10mL blood on liquid-handling systems.
Specifications
Content And Storage 15°C to 30°C
Quantity 4/Pk.