Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Integrin alpha 10 Alexa Fluor 488-conjugated Antibody, R&D Systems™
GREENER_CHOICESDP

Catalog No. FAB9757G100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB9757G100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB9757G100 Supplier R&D Systems Supplier No. FAB9757G100UG

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rat Monoclonal Antibody

Integrin alpha 10 Monoclonal specifically detects Integrin alpha 10 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen Integrin alpha 10
Applications Flow Cytometry
Classification Monoclonal
Clone 885501
Conjugate Alexa Fluor 488
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. NP_001289400.1 (Integrin alpha 10) and P09055 (Integrin beta 1)
Gene Alias integrin alpha-10, integrin, alpha 10, ITGA10, PRO827
Host Species Rat
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse Integrin alpha 10 beta 1 Phe23-Thr1119 (Integrin alpha 10) and Gln21-Asp728 (Integrin beta 1) Accession # NP_001289400.1 (Integrin alpha 10) and P09055 (Integrin beta 1)
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 8515
Test Specificity Detects mouse Integrin alpha 10 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse Intagrin alpha 2 beta 1 was observered.
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Supernatant
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.