Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Integrin alpha 4 beta 7 (LPAM-1) Monoclonal Antibody (DATK32 (DATK-32)), Super Bright™ 436, eBioscience™
GREENER_CHOICE

Catalog No. 62588782
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
62-588-782 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 62-588-782 Supplier Invitrogen™ Supplier No. 62588782
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rat Monoclonal Antibody

Description: The DATK32 monoclonal antibody binds to a combinatorial epitope of mouse LPAM-1 (alpha 4/ beta 7), which is a heterodimer of the the 154 kDa alpha 4 subunit and the 130 kDa beta 7 subunit. In mouse, the beta 7 subunit is found on the majority of mature lymphocytes and a small population of thymocytes and bone marrow cells. LPAM-1 is an integrin involved in the transendothelial migration of lymphocytes across high endothelial venules (HEV), and is know to bind several ligands including MAdCAM-1, VCAM-1 and fibronectin. Binding of the DATK32 monoclonal antibody has been shown to induce aggregation of the CD8+ T cell lymphoma TK1, and block LPAM-1-mediated cell adhesion. Applications Reported: This DATK32 antibody has been reported for use in flow cytometric analysis. Applications Tested: This DATK32 antibody has been tested by flow cytometric analysis of mouse bone marrow cells. This may be used at less than or equal to 1.0 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Super Bright 436 can be excited with the violet laser line (405 nm) and emits at 436 nm. We recommend using a 450/50 bandpass filter, or equivalent. Please make sure that your instrument is capable of detecting this fluor...

Integrin beta 7 associates with integrin alpha 4 (CD49d) to form the alpha 4 beta 7 integrin LPAM-1, expressed on intraepithelial lymphocytes. It also associates with alpha E (CD103) to form the alpha E beta 7 integrin HML-1, expressed on T cells adjacent to mucosal epithelium and intraepithelial lymphocytes. Ligands for LPAM-1 include VCAM-1 (CD106), MAdCAM-1 and fibronectin, while the ligand of HML-1 is E-cadherin (CD324). Integrin beta 7 plays an important role in the adhesion of leukocytes to endothelial cells promoting the transmigration of leukocytes to extravascular spaces during the inflammatory response.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Integrin alpha 4 beta 7 (LPAM-1)
Applications Flow Cytometry
Classification Monoclonal
Clone DATK32 (DATK-32)
Concentration 0.2 mg/mL
Conjugate Super Bright 436
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene Itgb7
Gene Accession No. P26011, Q00651
Gene Alias CD49 antigen-like family member D; CD49D; integrin alpha 4; integrin alpha-4; integrin alpha4B; Integrin alpha-IV; integrin beta 7; integrin beta-7; Integrin beta-P; Itga4; Itga4B; Itgb7; LPAM subunit alpha; Ly69; lymphocyte Peyer patch adhesion molecules subunit alpha; M290 IEL antigen; VLA; VLA 4; VLA-4 receptor, alpha 4 subunit; VLA4 subunit alpha; VLA-4 subunit alpha
Gene Symbols Itga4, Itgb7
Host Species Rat
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16401, 16421
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.