Learn More
Medchemexpress LLC Integrin alpha V bet 50ug | 50ug
SDP
Supplier:  Medchemexpress LLC HYP7772050UG
Encompass_Preferred
Integrin V beta 5 proteins specifically the -V (ITGAV) subunit are multifunctional receptors for ligands such as vitronectin and fibronectin and recognize RGD sequences ITGAV ITGB3 binds fractalkine and acts as a coreceptor in CX3CR1-dependent signaling Integrin alpha V beta 5 Protein Human (HEK293 His-Avi) is a recombinant protein dimer complex containing human-derived Integrin alpha V beta 5 protein expressed by HEK293 with C-Avi C-His labeled tag Integrin alpha V beta 5 Protein Human (HEK293 His-Avi) has molecular weight of 85-160 kDa
Catalog No. 50-001-87887
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.