Learn More
ACROBiosystems Integrin alpha V beta 5 1mg
Supplier:  ACROBiosystems IT5C52W31MG
Encompass_Preferred
Cynomolgus Integrin alpha V beta 5 (ITGAV&ITGB5) Heterodimer Protein His Tag&Tag Free (IT5-C52W3) is expressed from human 293 cells (HEK293). It contains AA Phe 31 - Pro 993 & Leu 25 - Asn 719 (Accession # A0A2K5WCD3-1 & A0A2K5UEB4).
Catalog No. 50-253-4545
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.