Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin beta 6 Antibody [Allophycocyanin], Novus Biologicals™
SDP

Catalog No. AF4155A
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF4155A 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AF4155A Supplier R&D Systems Supplier No. AF4155A

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Sheep Polyclonal Antibody

Integrin beta 6 Polyclonal antibody specifically detects Integrin beta 6 in Human samples. It is validated for Flow Cytometry,Western Blot,Immunocytochemistry,CyTOF

Specifications

Antigen Integrin beta 6
Applications Flow Cytometry, Western Blot, Immunocytochemistry, CyTOF
Classification Polyclonal
Conjugate APC
Formulation PBS
Gene Alias integrin beta-6, integrin, beta 6
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant human Integrin β6, Gly22-Asn707, Accession # P18564
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 3694
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.