Learn More
LI-COR Intercept (PBS) Blocking Buffer and Diluent Kit
SDP
Supplier:  LI-COR 92776003
Encompass
Try Intercept (PBS) Blocking Buffer and Intercept T20 (PBS) Antibody Diluent for improved results in your quantitative Western blots and other immunoassays. Intercept Blocking Buffers provide excellent blocking performance with low background and low variability.Intercept T20 Antibody Diluents improve the specificity of the primary and secondary antibodies, reducing off-target effects. They are preformulated with Tween 20. There’s no need to mix the diluent yourself, which saves you time and reduces potential variation.Kit Components: Intercept (PBS) Blocking Buffer, 1 x 500 mL; Intercept T20 (PBS) Antibody Diluent, 2 x 500 mLShake well before each use.Intercept (PBS) Blocking Buffer and Intercept T20 (PBS) Antibody Diluent can be used for many immunoassays and applications, including: Quantitative Western Blot, Chemiluminescent Western Blot, In-Cell Western™ Assay, In-Gel Western, Protein Array, Glycoprotein Detection
Catalog No. NC1698470
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.