Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Interferon alpha-10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. pipa597858
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597858 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597858 Supplier Invitrogen™ Supplier No. PA597858
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Interferon alpha-10 Polyclonal Antibody for Western Blot, IHC (P), ELISA

Produced by macrophages, IFN-alpha have antiviral activities. Interferon stimulates the production of two enzymes: a protein kinase and an oligoadenylate synthetase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Interferon alpha-10
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene IFNA10
Gene Accession No. P01566
Gene Alias IFN alpha; IFNA10; IFN-alpha-10; IFN-alphaC; Interferon; interferon alpha 10; interferon alpha-10; Interferon alpha-6L; interferon alpha-C; interferon, alpha 10; leIF C
Gene Symbols IFNA10
Host Species Rabbit
Immunogen Recombinant huamn Interferon alpha-10 protein (24-189AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 110296, 3446
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.