Learn More
Invitrogen™ Interferon alpha-2 Monoclonal Antibody (N27)

Description
The alpha interferons are involved in virus resistance in target cells for these viruses. They are known to block cell proliferation and to regulate MHC class I antigen expression. The IFNα family has over 20 genes and pseudogenes in two families (I and II), one with a mature length of 166aa and one of 172aa. Cells producing IFNα are lymphocytes, monocytes, macrophages and cell lines such as Namalwa and KGI. Bioassays for IFNα include cytopathic effect blocking, by viruses such as VSV, SFV and BMCV, on their target cells. A number of receptors for IFNα are now known and seem to be expressed on most cell types. N27 is specific for human IFNα2 and does not cross react with human IFNα1. N27 reacts with linear peptide 43aa-53aa, placing the epitope outside the immunodominant regions I and II.
Specifications
Specifications
| Antigen | Interferon alpha-2 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | N27 |
| Concentration | 100 μg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | IFNA2 |
| Gene Accession No. | P01563 |
| Gene Alias | alpha-2a interferon; Ifa2; IFNA; IFN-a; IFNA 2; Ifna2; IFNA2A; IFNA2B; IFNA2C; IFN-alpha 1b; IFN-alpha-1/13; IFN-alpha-2; IFN-alphaA; INFA2; Interferon; interferon alpha 1b; interferon alpha 2; interferon alpha 2a; interferon alpha 2b; interferon alpha A; interferon alpha family, gene 2; interferon alpha-2; Interferon alpha-A; interferon alpha-D; interferon, alpha 2; Interferon-alpha II; leIF A; leIF D; RP11-354P17.2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.