Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Interferon alpha-8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103429
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103429 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103429 Supplier Invitrogen™ Supplier No. PA5103429
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total IFNA8.

Interferon alpha-8 Produced by macrophages, IFN-alpha have antiviral activities. Interferon stimulates the production of two enzymes: a protein kinase and an oligoadenylate synthetase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Interferon alpha-8
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene IFNA8
Gene Accession No. P32881
Gene Alias IFNA8; IFN-alpha-8; IFN-alphaB; Interferon; interferon alpha 8; interferon alpha type 201; interferon alpha-8; Interferon alpha-B; interferon alpha-B'; interferon alpha-B2; interferon, alpha 8; leIF B; RP11-354P17.14-001
Gene Symbols IFNA8
Host Species Rabbit
Immunogen A synthesized peptide derived from human IFNA8(Accession P32881), corresponding to amino acid residues A76-E126.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3445
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.