Learn More
Creative Biomart Peginterferon beta-1a
Supplier:  Creative Biomart THP0023
Encompass_Preferred
The product is an interferon beta-1a to which a single, linear 20,000 dalton (Da) methoxy poly(ethyleneglycol)-O-2-methylpropionaldehyde (PEG) molecule is covalently attached to the alpha amino group of the N-terminal amino acid residue. The amino acid sequence of the recombinant interferon beta-1a is identical to that of the human interferon beta counterpart.
Catalog No. 50-208-4074
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.