Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Interferon Beta, Recombinant, Mouse, Full Length, 50 μg

Catalog No. 502704004
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-4004 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-4004 Supplier Abcam Supplier No. AB28239050UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant full length mouse Interferon beta protein expressed in HEK293 cells.

  • Protein Name: Interferon beta
  • Species: Mouse
  • Expressed In: HEK 293 cells
  • Sequence Length: Full Length (30 to 379 aa)
  • Purity: ≥95%
  • Endotoxin Level: ≤0.005 EU/μg
  • Form: Lyophilized
  • Applications: SDS-PAGE, HPLC, MS
  • Storage Buffer: pH 7.4; contains 10.26% Trehalose, 0.727% Dibasic monohydrogen potassium phosphate, and 0.428% Potassium phosphate monobasic
  • Animal Free: Yes

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Interferon Beta (Mouse)
Reactivity Mouse
Shipping Condition Ambient
Label or Dye Unconjugated
Content And Storage Store at ambient temperature.
Form Lyophilized
Buffer 10.26% trehalose, 0.727% dibasic monohydrogen potassium phosphate, 0.428% potassium phosphate monobasic, pH7.4
Formulation Lyophilized from pH 7.4 buffer containing 10.26% Trehalose, 0.727% Dibasic monohydrogen potassium phosphate, and 0.428% Potassium phosphate monobasic.
Quantity 50 μg
For Use With (Application) SDS-PAGE, HPLC, Mass Spec
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.